Gene Transcripts
- Gene Symbol
- Cuta
- Entrez Gene ID
- 67675
- Gene Name
- cutA divalent cation tolerance homolog (E. coli)
- Gene Aliases
- 0610039D01Rik, 1810022E02Rik, 1810060C03Rik, 2700094G22Rik, AI326454
- Chromosome Location
- Chr. 17: 26937972 - 26939815 on Build GRCm38
- UniGene ID
- Mm.439919
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026307.3 |
NP_080583.3
|
5, 6 |
454 |
|
NM_026948.3 |
NP_081224.1
|
4, 5 |
402 |
|
GenBank mRNA |
AK002828.1 |
- |
5, 6 |
398 |
|
AK003381.1 |
- |
4, 5 |
342 |
|
AK003647.1 |
- |
5, 6 |
396 |
|
AK004197.1 |
- |
5, 6 |
396 |
|
AK007580.1 |
- |
3, 4 |
531 |
|
AK007912.1 |
- |
4, 5 |
477 |
|
AK145185.1 |
- |
|
556 |
|
AK159336.1 |
- |
5, 6 |
401 |
|
AK199328.1 |
- |
2, 3 |
61 |
|
AK207031.1 |
- |
2, 3 |
61 |
|
AK207668.1 |
- |
1, 2 |
136 |
|
BC021463.1 |
- |
3, 4 |
516 |
|
BC024422.1 |
- |
5, 6 |
384 |
5005082 |
BC028531.1 |
- |
4, 5 |
493 |
|
CA323205.1 |
- |
4, 5 |
368 |
|
CK624897.1 |
- |
4, 5 |
379 |
|
View Full Product Details