Gene Transcripts
- Gene Symbol
- Dclk2
- Entrez Gene ID
- 70762
- Gene Name
- doublecortin-like kinase 2
- Gene Aliases
- 6330415M09Rik, AU044875, CL2, CLICK2, Click-II, Dcamkl2
- Chromosome Location
- Chr. 3: 86786150 - 86920884 on Build GRCm38
- UniGene ID
- Mm.44490
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195496.1 |
NP_001182425.1
|
14, 15 |
2639 |
|
NM_001195497.1 |
NP_001182426.1
|
13, 14 |
2591 |
|
NM_001195498.1 |
NP_001182427.1
|
14, 15 |
2642 |
|
NM_001195499.1 |
NP_001182428.1
|
14, 15 |
2639 |
|
NM_027539.5 |
NP_081815.3
|
13, 14 |
2594 |
|
GenBank mRNA |
AB198721.1 |
- |
14, 15 |
1996 |
|
AK049179.1 |
- |
13, 14 |
2043 |
|
AY968049.1 |
- |
14, 15 |
1927 |
|
AY968050.1 |
- |
14, 15 |
1930 |
|
AY968051.1 |
- |
14, 15 |
1927 |
|
BC052071.1 |
- |
6, 7 |
635 |
|
BC056921.1 |
- |
13, 14 |
2562 |
6822681 |
View Full Product Details