Gene Transcripts

Gene Symbol
Sept12
Entrez Gene ID
71089
Gene Name
septin 12
Gene Aliases
1700028G04Rik, 4933413B09Rik, Septin12
Chromosome Location
Chr. 16: 4986858 - 4997852 on Build GRCm38
UniGene ID
Mm.87382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027669.3 NP_081945.1 2 266
NM_029374.1 NP_083650.1 2 266
GenBank mRNA AK006458.1 - 2 204
AK016798.1 - 2 194
AK077091.1 - 2 267
BC115618.1 - 2 177 40102763
BC115619.1 - 2 177 40102765

View Full Product Details