Gene Transcripts

Gene Symbol
Immp1l
Entrez Gene ID
66541
Gene Name
IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae)
Gene Aliases
1500034J20Rik, 2610528O17Rik, AU042386
Chromosome Location
Chr. 2: 105904638 - 105965558 on Build GRCm38
UniGene ID
Mm.272253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028260.2 NP_082536.1 5 504
GenBank mRNA AK005356.1 - 5 505
AK009570.1 - 5 462
AK010075.1 - 5 462
AK011278.1 - 5 505
AK011382.1 - 5 505
AK011420.1 - 5 466
AK011466.1 - 5 477
AK011618.1 - 5 466
AK011872.1 - 5 468
AK012181.1 - 457
AK015978.1 - 5 475
AK078300.1 - 5 491
AK135670.1 - 5 464
BC008259.1 - 5 463 2649406
BC081433.1 - 4 347 6814816

View Full Product Details