Gene Transcripts

Gene Symbol
Lce1l
Entrez Gene ID
73730
Gene Name
late cornified envelope 1L
Gene Aliases
1110008K04Rik, AI415426
Chromosome Location
Chr. 3: 92849949 - 92851286 on Build GRCm38
UniGene ID
Mm.291773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028628.2 NP_082904.1 2 499
GenBank mRNA AK003585.1 - 2 500

View Full Product Details