Gene Transcripts

Gene Symbol
Atp11b
Entrez Gene ID
76295
Gene Name
ATPase, class VI, type 11B
Gene Aliases
1110019I14Rik, ATPIF, ATPIR, mKIAA0956
Chromosome Location
Chr. 3: 35754062 - 35856276 on Build GRCm38
UniGene ID
Mm.239416
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029570.3 NP_083846.2 29, 30 3700
GenBank mRNA AK003820.1 - 2, 3 601
AK129248.1 - 29, 30 3692
BC026741.1 - 3, 4 421
BC026986.1 - 14, 15 1769
BC076603.1 - 29, 30 3570 30629594
CF952078.1 - 2, 3 314

View Full Product Details