Gene Transcripts

Gene Symbol
Ms4a4c
Entrez Gene ID
64380
Gene Name
membrane-spanning 4-domains, subfamily A, member 4C
Gene Aliases
5830413L19Rik, AI449185, Ms4a9
Chromosome Location
Chr. 19: 11404717 - 11427246 on Build GRCm38
UniGene ID
Mm.440715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029499.3 NP_083775.1 7 1074
GenBank mRNA AB026045.1 - 7 1080
AF237914.2 - 7 1048
AK017928.1 - 7 1054
AK151045.1 - 7 1067
AK172650.1 - 7 1046
BC027496.1 - 7 1030 1383416

View Full Product Details