Gene Transcripts

Gene Symbol
Lrrc9
Entrez Gene ID
78257
Gene Name
leucine rich repeat containing 9
Gene Aliases
4921529O18Rik, 4930432K16Rik
Chromosome Location
Chr. 12: 72433940 - 72530174 on Build GRCm38
UniGene ID
Mm.266280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142728.1 NP_001136200.1 3 544
NM_001142729.1 NP_001136201.1 3 463
NM_030070.3 NP_084346.2 3 544
GenBank mRNA AK015292.1 - 3 462
AK019545.1 - 3 477
AK029802.1 - 3 545
AK031279.1 - 3 463
AK039037.1 - 3 354
AK082549.1 - 3 471

View Full Product Details