Gene Transcripts

Gene Symbol
Usp54
Entrez Gene ID
78787
Gene Name
ubiquitin specific peptidase 54
Gene Aliases
4930429G18Rik, AI115571, C030002J06Rik
Chromosome Location
Chr. 14: 20548912 - 20643129 on Build GRCm38
UniGene ID
Mm.301173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030180.2 NP_084456.2 21 5823
GenBank mRNA AK019588.1 - 4 2011
BC066166.1 - 4 2334
BC066175.1 - 4 2334
BC117844.1 - 4 1998
BC117845.1 - 4 1998

View Full Product Details