Gene Transcripts

Gene Symbol
Pcdhgb5
Entrez Gene ID
93702
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
Chromosome Location
Chr. 18: 37731154 - 37841870 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033577.1 NP_291055.1 1 740
GenBank mRNA AK156301.1 - 1 843
AY013807.1 - 1 740
BC057403.1 - 1 885

View Full Product Details