Gene Transcripts

Gene Symbol
Dmpk
Entrez Gene ID
13400
Gene Name
dystrophia myotonica-protein kinase
Gene Aliases
DM, DMK, Dm15, MDPK, MT-PK
Chromosome Location
Chr. 7: 19083834 - 19093821 on Build GRCm38
UniGene ID
Mm.6529
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190490.1 NP_001177419.1 2 357
NM_001190491.1 NP_001177420.1 2 357
NM_032418.2 NP_115794.1 2 357
GenBank mRNA BC024150.1 - 2 344
BC056615.1 - 2 336
BC075715.1 - 2 265 4989259

View Full Product Details