Gene Transcripts

Gene Symbol
Adam33
Entrez Gene ID
110751
Gene Name
a disintegrin and metallopeptidase domain 33
Gene Aliases
Adaml
Chromosome Location
Chr. 2: 131050817 - 131063814 on Build GRCm38
UniGene ID
Mm.108550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163529.1 NP_001157001.1 4 582
NM_033615.2 NP_291093.2 4 582
GenBank mRNA AB059632.1 - 4 582
AB059633.1 - 4 582
AF472524.1 - 4 314
AK053825.1 - 5 386
AY054984.1 - 4 282
AY382193.1 - 4 313
AY382194.1 - 4 313
BC138139.1 - 4 305 8861158

View Full Product Details