Gene Transcripts

Gene Symbol
Dqx1
Entrez Gene ID
93838
Gene Name
DEAQ RNA-dependent ATPase
Gene Aliases
2310066E11Rik
Chromosome Location
Chr. 6: 83057731 - 83067219 on Build GRCm38
UniGene ID
Mm.140332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033606.3 NP_291084.1 11 2213
GenBank mRNA AF318278.1 - 10 1857
BC048936.1 - 6 876
BC075680.1 - 11 2213 30612145

View Full Product Details