Gene Transcripts
- Gene Symbol
- Klra22
- Entrez Gene ID
- 93969
- Gene Name
- killer cell lectin-like receptor subfamily A, member 22
- Gene Aliases
- Ly49v
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333431
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053152.2 |
NP_444382.1
|
1, 2 |
256 |
|
GenBank mRNA |
AF288381.1 |
- |
1, 2 |
256 |
|
View Full Product Details