Gene Transcripts

Gene Symbol
Mcoln1
Entrez Gene ID
94178
Gene Name
mucolipin 1
Gene Aliases
2210015I05Rik, TRPML1, mucolipidin
Chromosome Location
Chr. 8: 3500442 - 3515232 on Build GRCm38
UniGene ID
Mm.8356
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053177.1 NP_444407.1 2 293
GenBank mRNA AF302009.1 - 2 290
AK028385.1 - 2 270
AK034471.1 - 2 304
AK038594.1 - 2 306
AK049606.1 - 2 294
BC005651.1 - 2 293 3257440
BC020377.1 - 1 143

View Full Product Details