Gene Transcripts

Gene Symbol
Ttyh2
Entrez Gene ID
117160
Gene Name
tweety homolog 2 (Drosophila)
Gene Aliases
1110001A03Rik
Chromosome Location
Chr. 11: 114675468 - 114720984 on Build GRCm38
UniGene ID
Mm.271934
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053273.2 NP_444503.2 4 518
GenBank mRNA AF329682.1 - 4 461
AK152172.1 - 4 470
AK154835.1 - 4 526
AK155166.1 - 4 519
AK156393.1 - 4 471
AK168408.1 - 4 471
BC060740.1 - 4 461 4158813
BY740853.1 - 4 498

View Full Product Details