Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267707.1 | NP_001254636.1 | 7 | 788 | |
NM_130861.3 | NP_570931.1 | 7 | 736 | ||
GenBank mRNA | AF240694.1 | - | 7 | 706 | |
AK029706.1 | - | 7 | 737 | ||
AK166446.1 | - | 7 | 765 |