Gene Transcripts

Gene Symbol
Nt5m
Entrez Gene ID
103850
Gene Name
5',3'-nucleotidase, mitochondrial
Gene Aliases
2010013E09Rik, AI846937, dNT-2
Chromosome Location
Chr. 11: 59847324 - 59876533 on Build GRCm38
UniGene ID
Mm.41761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134029.2 NP_598790.1 5 698
GenBank mRNA AK008226.1 - 666
AY061970.1 - 5 685
BC020084.1 - 5 698 3986525

View Full Product Details