Gene Transcripts

Gene Symbol
Vmn1r78
Entrez Gene ID
171242
Gene Name
vomeronasal 1 receptor 78
Gene Aliases
V1rg7
Chromosome Location
Chr. 7: 12152464 - 12153405 on Build GRCm38
UniGene ID
Mm.377198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134208.2 NP_598969.2 1 701
GenBank mRNA BC107194.1 - 1 701
BC107195.1 - 1 701 40051039

View Full Product Details