Gene Transcripts

Gene Symbol
Shkbp1
Entrez Gene ID
192192
Gene Name
Sh3kbp1 binding protein 1
Gene Aliases
B930062H15Rik, Sb1
Chromosome Location
Chr. 7: 27342128 - 27356015 on Build GRCm38
UniGene ID
Mm.21297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138676.2 NP_619617.2 6 398
GenBank mRNA AF246218.1 - 6 404
AK047434.1 - 4 471
AK154779.1 - 6 387
AK160641.1 - 5 470
AK166328.1 - 5 343
BB847999.1 - 6 401
BC061477.1 - 6 351 6336438

View Full Product Details