Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144535.4 | NP_653118.3 | 2 | 502 | |
GenBank mRNA | AK016540.1 | - | 2 | 449 | |
AK028927.1 | - | 2 | 455 | ||
AK030928.1 | - | 2 | 429 | ||
AK031527.1 | - | 2 | 901 | ||
AK036454.1 | - | 2 | 427 | ||
AK036465.1 | - | 2 | 434 | ||
AK041400.1 | - | 2 | 455 | ||
AK042544.1 | - | 2 | 424 | ||
AK051372.1 | - | 2 | 449 | ||
AK154166.1 | - | 2 | 455 | ||
AK164148.1 | - | 2 | 273 | ||
AK170232.1 | - | 2 | 279 | ||
AK211895.1 | - | 2 | 179 | ||
BC023094.1 | - | 2 | 502 | 4017596 |