Gene Transcripts

Gene Symbol
Ankrd54
Entrez Gene ID
223690
Gene Name
ankyrin repeat domain 54
Gene Aliases
C730048E16Rik, EST1068184, EST475269, Liar
Chromosome Location
Chr. 15: 79053094 - 79062958 on Build GRCm38
UniGene ID
Mm.24262
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144849.1 NP_659098.1 1, 2 385
GenBank mRNA AK050440.1 - 1, 2 412
AK084500.1 - 1, 2 420
AK133785.1 - 1, 2 409
AK143625.1 - 1, 2 412
BC014743.1 - 1, 2 385 4503319
DQ884878.1 - 1, 2 413

View Full Product Details