Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301705.1 | NP_001288634.1 | 3, 4 | 376 | |
NM_144899.3 | NP_659148.2 | 3, 4 | 258 | ||
GenBank mRNA | AK155481.1 | - | 3, 4 | 306 | |
AY158701.1 | - | 3, 4 | 199 | ||
BC117925.1 | - | 2, 3 | 171 | 40097608 | |
BC117926.1 | - | 2, 3 | 172 | ||
BY196249.1 | - | 3, 4 | 339 | ||
BY204619.1 | - | 3, 4 | 355 |