Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145211.2 | NP_660212.2 | 6, 7 | 1128 | |
GenBank mRNA | AK149613.1 | - | 6, 7 | 1089 | |
BC013715.1 | - | 6, 7 | 1110 | 3257027 | |
BC057878.1 | - | 6, 7 | 1087 | 5348858 | |
M33863.1 | - | 1114 | |||
X04958.1 | - | 6, 7 | 1114 |