Gene Transcripts

Gene Symbol
Uts2r
Entrez Gene ID
217369
Gene Name
urotensin 2 receptor
Gene Aliases
Gpr14, UR-2-R, UTR, UTR2
Chromosome Location
Chr. 11: 121160271 - 121161973 on Build GRCm38
UniGene ID
Mm.20122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145440.1 NP_663415.1 1 1469
GenBank mRNA AF441863.1 - 1 1469
BC125312.1 - 1 1448 40129827
BC125318.1 - 1 1448 40129833

View Full Product Details