Gene Transcripts

Gene Symbol
Tiprl
Entrez Gene ID
226591
Gene Name
TIP41, TOR signalling pathway regulator-like (S. cerevisiae)
Gene Aliases
1810011K17Rik
Chromosome Location
Chr. 1: 165212286 - 165282478 on Build GRCm38
UniGene ID
Mm.21520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145513.4 NP_663488.1 7 3936
GenBank mRNA AK049784.1 - 7 3923

View Full Product Details