Gene Transcripts
- Gene Symbol
- Evi2b
- Entrez Gene ID
- 216984
- Gene Name
- ecotropic viral integration site 2b
- Gene Aliases
- Evi-2, Evi2
- Chromosome Location
- Chr. 11: 79513385 - 79523762 on Build GRCm38
- UniGene ID
- Mm.439665
- Species
- Mus musculus
- Gene Symbol
- Gm21975
- Entrez Gene ID
- 101488212
- Gene Name
- predicted gene 21975
- Gene Aliases
- Evi2a-evi2b
- Chromosome Location
- Chr. 11: 79513385 - 79530609 on Build GRCm38
- UniGene ID
- Mm.439665
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146023.4 |
NP_666135.1
|
4 |
763 |
|
View Full Product Details