Gene Transcripts

Gene Symbol
Ppp4r1
Entrez Gene ID
70351
Gene Name
protein phosphatase 4, regulatory subunit 1
Gene Aliases
3110001J10Rik, Pp4r1
Chromosome Location
Chr. 17: 65782398 - 65841926 on Build GRCm38
UniGene ID
Mm.48686
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114131.1 NP_001107603.1 11 1290
NM_146081.2 NP_666193.2 11 1341
GenBank mRNA AK133969.1 - 11 1212
AK166418.1 - 11 1248
BC026489.1 - 3 314 4504305
BC029761.1 - 11 1276 3989055

View Full Product Details