Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000716.3 | NP_000707.1 | 4 | 755 | |
NM_001017364.1 | NP_001017364.1 | 4 | 752 | ||
NM_001017365.2 | NP_001017365.1 | 5 | 617 | ||
NM_001017366.2 | NP_001017366.1 | 5 | 614 | ||
NM_001017367.1 | NP_001017367.1 | 5 | 591 | ||
XM_005273254.4 | XP_005273311.1 | 6 | 896 | ||
GenBank mRNA | AK315137.1 | - | 5 | 859 | |
BC005378.1 | - | 4 | 752 | 3934453 | |
L11245.1 | - | 4 | 740 | ||
M29964.1 | - | 4 | 490 |