Gene Transcripts

Gene Symbol
Lrrk1
Entrez Gene ID
233328
Gene Name
leucine-rich repeat kinase 1
Gene Aliases
AW319595, C230002E15Rik, D130026O16Rik, mKIAA1790
Chromosome Location
Chr. 7: 66258745 - 66388341 on Build GRCm38
UniGene ID
Mm.33284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146191.3 NP_666303.3 34 6220
GenBank mRNA AK040153.1 - 14 2902
AK076432.1 - 7 1547
AK081849.1 - 4 1213
AK129441.1 - 31 5633
AK147469.1 - 34 6206
AK154395.1 - 8 1739
BC027199.1 - 4 1088
BC066159.1 - 3 742
BC072664.1 - 24 3688 30543489
BC080819.1 - 3 847

View Full Product Details