Gene Transcripts

Gene Symbol
Foxn4
Entrez Gene ID
116810
Gene Name
forkhead box N4
Gene Aliases
Chromosome Location
Chr. 5: 114254164 - 114274598 on Build GRCm38
UniGene ID
Mm.17709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148935.2 NP_683737.2 5 499
GenBank mRNA AF323488.1 - 5 507
AK081735.1 - 3 204
AY039039.1 - 5 545
BC092242.1 - 5 488 30698162
BC130222.1 - 5 500 40141600

View Full Product Details