Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190801.1 | NP_001177730.1 | 3 | 285 | |
NM_014264.4 | NP_055079.3 | 3 | 462 | ||
XM_005262701.2 | XP_005262758.1 | 3 | 424 | ||
XM_017007662.1 | XP_016863151.1 | 3 | 621 | ||
XM_017007663.1 | XP_016863152.1 | 3 | 622 | ||
GenBank mRNA | AB006972.1 | - | 3 | 376 | |
AB741646.1 | - | 3 | 188 | ||
AK302858.1 | - | 3 | 285 | ||
AK314238.1 | - | 3 | 430 | ||
BC036023.1 | - | 3 | 430 | 5273226 | |
BP328161.1 | - | 3 | 434 | ||
Y13115.1 | - | 3 | 328 |