Gene Transcripts
- Gene Symbol
- Myl6b
- Entrez Gene ID
- 216459
- Gene Name
- myosin, light polypeptide 6B
- Gene Aliases
- 5730437E04Rik, BC037527, MLC1SA
- Chromosome Location
- Chr. 10: 128494157 - 128498685 on Build GRCm38
- UniGene ID
- Mm.291688
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172259.1 |
NP_758463.1
|
3, 4 |
310 |
|
GenBank mRNA |
BC037527.1 |
- |
3, 4 |
310 |
|
View Full Product Details