Gene Transcripts
- Gene Symbol
- Astl
- Entrez Gene ID
- 215095
- Gene Name
- astacin-like metalloendopeptidase (M12 family)
- Gene Aliases
- 7420700F21, C87576, Sas1b
- Chromosome Location
- Chr. 2: 127338639 - 127357655 on Build GRCm38
- UniGene ID
- Mm.31313
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291003.1 |
NP_001277932.1
|
2, 3 |
250 |
|
NM_172539.3 |
NP_766127.1
|
2, 3 |
381 |
|
GenBank mRNA |
AJ537599.2 |
- |
2, 3 |
189 |
|
AK033037.1 |
- |
2, 3 |
381 |
|
BC064729.1 |
- |
2, 3 |
242 |
30476764 |
DV649054.1 |
- |
2, 3 |
262 |
|
FJ187790.1 |
- |
2, 3 |
189 |
|
FJ187791.1 |
- |
1, 2 |
126 |
|
FJ187792.1 |
- |
1, 2 |
126 |
|
FJ187793.1 |
- |
2, 3 |
189 |
|
FJ187794.1 |
- |
1, 2 |
126 |
|
FJ187795.1 |
- |
2, 3 |
189 |
|
View Full Product Details