Gene Transcripts

Gene Symbol
Epc2
Entrez Gene ID
227867
Gene Name
enhancer of polycomb homolog 2 (Drosophila)
Gene Aliases
5830499L14, D2Ertd694e
Chromosome Location
Chr. 2: 49451486 - 49551942 on Build GRCm38
UniGene ID
Mm.29167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172663.4 NP_766251.3 1, 2 469
GenBank mRNA AK031025.1 - 1, 2 442
AK047947.1 - 1, 2 137
BB620197.1 - 1, 2 431

View Full Product Details