Gene Transcripts

Gene Symbol
Epc2
Entrez Gene ID
227867
Gene Name
enhancer of polycomb homolog 2 (Drosophila)
Gene Aliases
5830499L14, D2Ertd694e
Chromosome Location
Chr. 2: 49451486 - 49551942 on Build GRCm38
UniGene ID
Mm.29167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172663.4 NP_766251.3 14 3601
GenBank mRNA AK031025.1 - 14 3574
AK141073.1 - 2892
BC048785.1 - 13 3040

View Full Product Details