Gene Transcripts

Gene Symbol
Mtmr10
Entrez Gene ID
233315
Gene Name
myotubularin related protein 10
Gene Aliases
BB128963, C79734
Chromosome Location
Chr. 7: 64287670 - 64340806 on Build GRCm38
UniGene ID
Mm.214959
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172742.2 NP_766330.2 15 1817
GenBank mRNA AK028584.1 - 3 442
BC041672.1 - 15 1789
BC055074.1 - 13 1481 30052208
BC058196.1 - 8 919
CB600359.1 - 3 488

View Full Product Details