Gene Transcripts

Gene Symbol
Klhl4
Entrez Gene ID
237010
Gene Name
kelch-like 4
Gene Aliases
B130014A11, C130018J01Rik
Chromosome Location
Chr. X: 114474333 - 114561129 on Build GRCm38
UniGene ID
Mm.132583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290477.1 NP_001277406.1 11 2367
NM_001290478.1 NP_001277407.1 11 2286
NM_172781.2 NP_766369.2 11 2385
GenBank mRNA AK044930.1 - 11 2017
AK081448.1 - 11 2369
AK081937.1 - 11 2198
AK085099.1 - 11 2368
AK161786.1 - 11 2260
BC046395.1 - 5 945
BC057137.1 - 11 2116
BC141251.1 - 11 2110 9055863

View Full Product Details