Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130.5 | NP_001121.2 | 5 | 432 | |
NM_198969.1 | NP_945320.1 | 5 | 499 | ||
NM_198970.1 | NP_945321.1 | 5 | 432 | ||
XM_006722664.1 | XP_006722727.1 | 5 | 640 | ||
GenBank mRNA | AF072902.1 | - | 5 | 335 | |
AK094591.1 | - | 5 | 641 | ||
AK095154.1 | - | 2 | 484 | ||
AK314713.1 | - | 5 | 338 | ||
BC113735.1 | - | 5 | 486 | 8322787 | |
BC113737.1 | - | 5 | 486 | 8322789 | |
BG696926.1 | - | 1 | 16 | ||
BG913049.1 | - | 5 | 419 | ||
BM550097.1 | - | 389 | |||
BM908356.1 | - | 460 | |||
U04241.1 | - | 5 | 299 | ||
X73357.1 | - | 5 | 253 | ||
X73358.1 | - | 4 | 223 |