Gene Transcripts

Gene Symbol
Agtr1b
Entrez Gene ID
11608
Gene Name
angiotensin II receptor, type 1b
Gene Aliases
AT1B, AT2R1B, Agtr-1b, Angtr-1b
Chromosome Location
Chr. 3: 20314473 - 20367177 on Build GRCm38
UniGene ID
Mm.134863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175086.3 NP_780295.2 2 859
GenBank mRNA AK087228.1 - 2 859

View Full Product Details