Gene Transcripts

Gene Symbol
Clspn
Entrez Gene ID
269582
Gene Name
claspin
Gene Aliases
B130025E01, C85083, E130314M08Rik
Chromosome Location
Chr. 4: 126556951 - 126594533 on Build GRCm38
UniGene ID
Mm.243962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175554.4 NP_780763.3 2 238
GenBank mRNA AK045076.1 - 2 234
AK051966.1 - 2 213
AK053847.1 - 2 252
AK135394.1 - 2 232
BC050848.1 - 2 193 6507692
CF535627.1 - 2 238

View Full Product Details