Gene Transcripts

Gene Symbol
Rab39
Entrez Gene ID
270160
Gene Name
RAB39, member RAS oncogene family
Gene Aliases
C230094F14Rik, Rab39a
Chromosome Location
Chr. 9: 53684110 - 53706232 on Build GRCm38
UniGene ID
Mm.290285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175562.3 NP_780771.1 2 390
GenBank mRNA AB232635.1 - 2 273
AK049038.1 - 2 390
AK053487.1 - 2 387
BC049787.1 - 2 370 6511404

View Full Product Details