Gene Transcripts

Gene Symbol
CST2
Entrez Gene ID
1470
Gene Name
cystatin SA
Gene Aliases
Chromosome Location
Chr. 20: 23823767 - 23826675 on Build GRCh38
UniGene ID
Hs.669305
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322.2 NP_001313.1 1 95
GenBank mRNA BC062679.1 - 1 153

View Full Product Details