Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176838.2 | NP_789808.1 | 4, 5 | 770 | |
GenBank mRNA | AK088755.1 | - | 4, 5 | 639 | |
AK168488.1 | - | 4, 5 | 618 | ||
BC031444.1 | - | 4, 5 | 770 | 4222237 | |
BC096581.1 | - | 4, 5 | 610 |