Gene Transcripts

Gene Symbol
Ablim3
Entrez Gene ID
319713
Gene Name
actin binding LIM protein family, member 3
Gene Aliases
AW990719, D930036B08Rik
Chromosome Location
Chr. 18: 61799392 - 61911857 on Build GRCm38
UniGene ID
Mm.329478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164491.1 NP_001157963.1 2 194
NM_198649.3 NP_941051.2 1 513
GenBank mRNA AK082380.1 - 1 512
AK086538.1 - 1 293
AK173040.1 - 1 71
BC060275.1 - 2 165 6822626
BC094229.1 - 1 487 30534236

View Full Product Details