Gene Transcripts

Gene Symbol
Samd5
Entrez Gene ID
320825
Gene Name
sterile alpha motif domain containing 5
Gene Aliases
E130306M17Rik
Chromosome Location
Chr. 10: 9627259 - 9675208 on Build GRCm38
UniGene ID
Mm.101115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177271.3 NP_796245.2 2 857
GenBank mRNA AK045645.1 - 2 409
AK087498.1 - 2 859
AK132444.1 - 2 831
BC145672.1 - 2 631 40130975
BC145674.1 - 2 631 40130977

View Full Product Details