Gene Transcripts
- Gene Symbol
- Pisd
- Entrez Gene ID
- 320951
- Gene Name
- phosphatidylserine decarboxylase
- Gene Aliases
- 9030221M09Rik
- Chromosome Location
- Chr. 5: 32736303 - 32785636 on Build GRCm38
- UniGene ID
- Mm.273765
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177298.3 |
NP_796272.2
|
2, 3 |
157 |
|
GenBank mRNA |
AK033483.1 |
- |
2, 3 |
149 |
|
AK034656.1 |
- |
2, 3 |
159 |
|
AK162028.1 |
- |
2, 3 |
150 |
|
AK162737.1 |
- |
2, 3 |
158 |
|
AK167554.1 |
- |
1, 2 |
99 |
|
BC070408.1 |
- |
2, 3 |
156 |
6809528 |
BC094594.1 |
- |
2, 3 |
148 |
5707749 |
BC116695.1 |
- |
2, 3 |
159 |
40105525 |
BC117509.1 |
- |
2, 3 |
159 |
40105529 |
CX201846.1 |
- |
2, 3 |
172 |
|
View Full Product Details