Gene Transcripts

Gene Symbol
Vangl1
Entrez Gene ID
229658
Gene Name
vang-like 1 (van gogh, Drosophila)
Gene Aliases
Kitenin, Lpp2, Stb2, Stbm
Chromosome Location
Chr. 3: 102155899 - 102205011 on Build GRCm38
UniGene ID
Mm.118004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177545.5 NP_808213.2 3 408
GenBank mRNA AF481860.1 - 3 324
AK135230.1 - 3 369
AK167743.1 - 3 290
BC138519.1 - 2 150 8861539
BC138520.1 - 2 150 8861540
BY729182.1 - 3 408

View Full Product Details