Gene Transcripts

Gene Symbol
Atmin
Entrez Gene ID
234776
Gene Name
ATM interactor
Gene Aliases
Asciz, gpg6, mKIAA0431
Chromosome Location
Chr. 8: 116943386 - 116960445 on Build GRCm38
UniGene ID
Mm.259736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177700.4 NP_808368.3 4 930
GenBank mRNA AK087334.1 - 1 241
AK122281.1 - 4 812
AX775769.1 - 4 930
BC058974.1 - 3 475
BC060631.1 - 4 915 6847850

View Full Product Details