Gene Transcripts
- Gene Symbol
- Vmn2r62
- Entrez Gene ID
- 546964
- Gene Name
- vomeronasal 2, receptor 62
- Gene Aliases
- Chromosome Location
- Chr. 7: 42764438 - 42793496 on Build GRCm38
- UniGene ID
- Mm.469898
- Species
- Mus musculus
- Gene Symbol
- Gm9268
- Entrez Gene ID
- 668612
- Gene Name
- predicted gene 9268
- Gene Aliases
- EG668612, Vmn2r64
- Chromosome Location
- Chr. 7: 43018798 - 43048106 on Build GRCm38
- UniGene ID
- Mm.469896
- Species
- Mus musculus
- Gene Symbol
- Vmn2r57
- Entrez Gene ID
- 269902
- Gene Name
- vomeronasal 2, receptor 57
- Gene Aliases
- D430014M15, EG269902
- Chromosome Location
- Chr. 7: 41399732 - 41448641 on Build GRCm38
- UniGene ID
- Mm.483785
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177764.4 |
NP_808432.2
|
6 |
1868 |
|
- Gene Symbol
- Vmn2r63
- Entrez Gene ID
- 435975
- Gene Name
- vomeronasal 2, receptor 63
- Gene Aliases
- EG435975
- Chromosome Location
- Chr. 7: 42903251 - 42933789 on Build GRCm38
- UniGene ID
- Mm.469897
- Species
- Mus musculus
View Full Product Details